missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N^w-Nitro-L-arginine methyl ester hydrochloride, 98%
CAS: 51298-62-5 | C7H16ClN5O4 | 269.69 g/mol
$198.47 - $445.68
Chemical Identifiers
| CAS | 51298-62-5 |
|---|---|
| Molecular Formula | C7H16ClN5O4 |
| Molecular Weight (g/mol) | 269.69 |
| MDL Number | MFCD00039052,MFCD00133613 |
| InChI Key | QBNXAGZYLSRPJK-JEDNCBNOSA-N |
| Synonym | h-arg no2-ome.hcl, lname hydrochloride, l-name hydrochloride, ng-nitro-l-arginine methyl ester hydrochloride, nomega-nitro-l-arginine methyl ester hydrochloride, s-methyl 2-amino-5-3-nitroguanidino pentanoate hydrochloride, l-name, hcl, ng-no2-l-arg-ome, nomega-no2-l-arg-ome |
| PubChem CID | 135193 |
| IUPAC Name | methyl (2S)-2-amino-5-[[amino(nitramido)methylidene]amino]pentanoate;hydrochloride |
| SMILES | [H+].[Cl-].COC(=O)[C@@H](N)CCCN=C(N)N[N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6366606
|
Thermo Scientific Chemicals
H6366606 |
5 g |
Each for $198.47
|
|
|||||
|
AAH6366614
|
Thermo Scientific Chemicals
H6366614 |
25 g |
Each for $445.68
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 51298-62-5 | |
| 269.69 | |
| QBNXAGZYLSRPJK-JEDNCBNOSA-N | |
| 135193 | |
| [H+].[Cl-].COC(=O)[C@@H](N)CCCN=C(N)N[N+]([O-])=O |
| C7H16ClN5O4 | |
| MFCD00039052,MFCD00133613 | |
| h-arg no2-ome.hcl, lname hydrochloride, l-name hydrochloride, ng-nitro-l-arginine methyl ester hydrochloride, nomega-nitro-l-arginine methyl ester hydrochloride, s-methyl 2-amino-5-3-nitroguanidino pentanoate hydrochloride, l-name, hcl, ng-no2-l-arg-ome, nomega-no2-l-arg-ome | |
| methyl (2S)-2-amino-5-[[amino(nitramido)methylidene]amino]pentanoate;hydrochloride |
Specifications
| 159°C to 161°C | |
| White | |
| MFCD00039052,MFCD00133613 | |
| h-arg no2-ome.hcl, lname hydrochloride, l-name hydrochloride, ng-nitro-l-arginine methyl ester hydrochloride, nomega-nitro-l-arginine methyl ester hydrochloride, s-methyl 2-amino-5-3-nitroguanidino pentanoate hydrochloride, l-name, hcl, ng-no2-l-arg-ome, nomega-no2-l-arg-ome | |
| [H+].[Cl-].COC(=O)[C@@H](N)CCCN=C(N)N[N+]([O-])=O | |
| 5 g | |
| 135193 | |
| 98% | |
| Nω-Nitro-L-arginine methyl ester hydrochloride |
| 51298-62-5 | |
| C7H16ClN5O4 | |
| 3744166 | |
| QBNXAGZYLSRPJK-JEDNCBNOSA-N | |
| methyl (2S)-2-amino-5-[[amino(nitramido)methylidene]amino]pentanoate;hydrochloride | |
| 269.69 | |
| 269.69 | |
| Powder |
Safety and Handling
EINECSNumber : 257-116-1
RTECSNumber : RM2982570
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only