missing translation for 'onlineSavingsMsg'
Learn More

N^w-Nitro-L-arginine methyl ester hydrochloride, 98%

Catalog No. AAH6366614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6366614 25 g
AAH6366606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6366614 Supplier Thermo Scientific Chemicals Supplier No. H6366614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51298-62-5 | C7H16ClN5O4 | 269.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51298-62-5
Molecular Formula C7H16ClN5O4
Molecular Weight (g/mol) 269.69
MDL Number MFCD00039052,MFCD00133613
InChI Key QBNXAGZYLSRPJK-JEDNCBNOSA-N
Synonym h-arg no2-ome.hcl, lname hydrochloride, l-name hydrochloride, ng-nitro-l-arginine methyl ester hydrochloride, nomega-nitro-l-arginine methyl ester hydrochloride, s-methyl 2-amino-5-3-nitroguanidino pentanoate hydrochloride, l-name, hcl, ng-no2-l-arg-ome, nomega-no2-l-arg-ome
PubChem CID 135193
IUPAC Name methyl (2S)-2-amino-5-[[amino(nitramido)methylidene]amino]pentanoate;hydrochloride
SMILES [H+].[Cl-].COC(=O)[C@@H](N)CCCN=C(N)N[N+]([O-])=O

Specifications

Melting Point 159°C to 161°C
Color White
Beilstein 3744166
Quantity 25 g
Formula Weight 269.69
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Nω-Nitro-L-arginine methyl ester hydrochloride
EINECSNumber 257-116-1
RTECSNumber RM2982570
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only