missing translation for 'onlineSavingsMsg'
Learn More

N-Vinylphthalimide 98.0+%, TCI America™

Catalog No. V00655G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V00655G 5 g
V006525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V00655G Supplier TCI America Supplier No. V00655G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3485-84-5
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.17
MDL Number MFCD00078446
InChI Key IGDLZDCWMRPMGL-UHFFFAOYSA-N
Synonym n-vinylphthalimide, 2-vinylisoindoline-1,3-dione, phthalimide, n-vinyl, 1h-isoindole-1,3 2h-dione, 2-ethenyl, 2-vinyl-1h-isoindole-1,3 2h-dione, 2-ethenyl-1h-isoindole-1,3 2h-dione, 2-ethenyl-1,3-dihydro-1,3-dioxoisoindole, 2-ethenyl-2,3-dihydro-1h-isoindole-1,3-dione, 2-vinylbenzo c azolidine-1,3-dione, n-vinylphtalimide
PubChem CID 77035
IUPAC Name 2-ethenyl-2,3-dihydro-1H-isoindole-1,3-dione
SMILES C=CN1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 83°C
Boiling Point 151°C
Quantity 5 g
Formula Weight 173.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Vinylphthalimide