missing translation for 'onlineSavingsMsg'
Learn More

N-Vinylformamide (stabilized with BHT) 96.0+%, TCI America™

Catalog No. p-7137924
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V010625G 25 g
V0106500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V010625G Supplier TCI America Supplier No. V010625G

Chemical Identifiers

CAS 13162-05-5
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.079
MDL Number MFCD00081206
InChI Key ZQXSMRAEXCEDJD-UHFFFAOYSA-N
PubChem CID 83191
IUPAC Name N-ethenylformamide
SMILES C=CNC=O

Specifications

Melting Point -16°C
Boiling Point 87°C
Quantity 25 g
Formula Weight 71.08
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Vinylformamide (stabilized with BHT)
EINECSNumber (2)-3501
TSCA Yes
Recommended Storage Refrigerator