missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Vinylformamide (stabilized with BHT) 96.0+%, TCI America™
Specifications
Specifications
| Melting Point | -16°C |
| Boiling Point | 87°C |
| Quantity | 500 g |
| Formula Weight | 71.08 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | N-Vinylformamide (stabilized with BHT) |
Safety and Handling
| EINECSNumber | (2)-3501 |
| TSCA | Yes |
| Recommended Storage | Refrigerator |