missing translation for 'onlineSavingsMsg'
Learn More

N-Vinyl-2-pyrrolidone, 99%, stabilized with Kerobit(R), Thermo Scientific Chemicals

Catalog No. p-4510659
Change view
Click to view available options
Quantity:
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC354730025 2.5 L Glass Bottle
AC354730100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC354730025 Supplier Thermo Scientific Chemicals Supplier No. 354730025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 88-12-0
Molecular Formula C6H9NO
Molecular Weight (g/mol) 111.14
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
Synonym n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
PubChem CID 6917
ChEBI CHEBI:82551
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES C=CN1CCCC1=O

Specifications

Melting Point 13.0°C to 14.0°C
Color Colorless to Yellow
Density 1.0430g/mL
Boiling Point 218.0°C
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.5119 to 1.5139
Quantity 2.5 L
Merck Index 15,7814
Specific Gravity 1.043
Solubility Information Solubility in water: 52.1g/L (25°C). Other solubilities: miscible with acetone,diethyl ether,ethanol,,toluene,benzene,very good soluble in organic solvents
Stabilizer 10ppm kerobit
Viscosity 2.4 mPa.s (20°C)
Formula Weight 111.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N-Vinyl-2-pyrrolidone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 201-800-4
Recommended Storage Product may darken on storage

RUO – Research Use Only