missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Valerylglycine Analytical Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 24003-66-5 |
| Molecular Formula | C7H13NO3 |
| Linear Formula | C7H13NO3 |
| Quantity | 10 mg |
| MDL Number | MFCD01910091 |
| UN Number | NONH for all modes of transport |
| Synonym | 2-(Pentanoylamino)acetic acid; N-(1-Oxopentyl)glycine; N-Pentanoylglycine; Valeryl glycine |
| Formula Weight | 159.18 |
| Percent Purity | ≥98% (HPLC) |