missing translation for 'onlineSavingsMsg'
Learn More

n-Undecane, 99.7%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0215823190

 View more versions of this product

Catalog No. ICN15823190

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

n-Undecane
Colorless
0.997
MFCD00008959
n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und
CCCCCCCCCCC
156.31
CHEBI:46342
Liquid
1120-21-4
0.744 at 25°C (water = 1)
C11H24
500 mL
RSJKGSCJYJTIGS-UHFFFAOYSA-N
undecane
14257
156.3

Chemical Identifiers

1120-21-4
156.31
RSJKGSCJYJTIGS-UHFFFAOYSA-N
14257
undecane
C11H24
MFCD00008959
n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und
CHEBI:46342
CCCCCCCCCCC
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions