missing translation for 'onlineSavingsMsg'
Learn More

N-Tris(hydroxymethyl)methyl-3-aminopropanesulfonic Acid 98.0+%, TCI America™

Catalog No. T097425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T097425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T097425G Supplier TCI America Supplier No. T097425G

[Good's buffer component for biological research]

Chemical Identifiers

CAS 29915-38-6
Molecular Formula C7H17NO6S
Molecular Weight (g/mol) 243.27
MDL Number MFCD00007538
InChI Key YNLCVAQJIKOXER-UHFFFAOYSA-N
Synonym taps, 3-1,3-dihydroxy-2-hydroxymethyl propan-2-yl amino propane-1-sulfonic acid, unii-y5dc3in066, n-tris hydroxymethyl methyl-3-aminopropanesulfonic acid, 2-hydroxy-1,1-bis hydroxymethyl ethyl amino-1-propanesulfonic acid, 3-2-hydroxy-1,1-bis hydroxymethyl ethyl amino-1-propanesulfonic acid, taps buffer, 1-propanesulfonic acid, 3-2-hydroxy-1,1-bis hydroxymethyl ethyl amino
PubChem CID 121591
IUPAC Name 3-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}propane-1-sulfonic acid
SMILES OCC(CO)(CO)NCCCS(O)(=O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 243.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Tris(hydroxymethyl)methyl-3-aminopropanesulfonic Acid [Good′s buffer component for biologica