missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Tris(hydroxymethyl)methyl-2-aminoethanesulfonic Acid 98.0+%, TCI America™
[Good's buffer component for biological research]
$233.00 - $1741.03
Chemical Identifiers
| CAS | 7365-44-8 |
|---|---|
| Molecular Formula | C6H15NO6S |
| Molecular Weight (g/mol) | 229.247 |
| MDL Number | MFCD00007532 |
| InChI Key | JOCBASBOOFNAJA-UHFFFAOYSA-N |
| Synonym | tes, n-tris hydroxymethyl methyl-2-aminoethanesulfonic acid, tes buffer, tes free acid, ethanesulfonic acid, 2-2-hydroxy-1,1-bis hydroxymethyl ethyl amino, 2-1,3-dihydroxy-2-hydroxymethyl propan-2-yl amino ethanesulfonic acid, 2-2-hydroxy-1,1-dihydroxymethyl-ethylamino-ethanesulfonic acid, tris hydroxymethyl methyl-2-aminomethane sulfonic acid, 2-tris hydroxymethyl methylamino ethane-1-sulphonic acid, n-tri hydroxymethyl methyltaurine |
| PubChem CID | 81831 |
| ChEBI | CHEBI:44356 |
| IUPAC Name | 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonic acid |
| SMILES | C(CS(=O)(=O)O)NC(CO)(CO)CO |
Chemical Identifiers
| 7365-44-8 | |
| 229.247 | |
| JOCBASBOOFNAJA-UHFFFAOYSA-N | |
| 81831 | |
| 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonic acid |
| C6H15NO6S | |
| MFCD00007532 | |
| tes, n-tris hydroxymethyl methyl-2-aminoethanesulfonic acid, tes buffer, tes free acid, ethanesulfonic acid, 2-2-hydroxy-1,1-bis hydroxymethyl ethyl amino, 2-1,3-dihydroxy-2-hydroxymethyl propan-2-yl amino ethanesulfonic acid, 2-2-hydroxy-1,1-dihydroxymethyl-ethylamino-ethanesulfonic acid, tris hydroxymethyl methyl-2-aminomethane sulfonic acid, 2-tris hydroxymethyl methylamino ethane-1-sulphonic acid, n-tri hydroxymethyl methyltaurine | |
| CHEBI:44356 | |
| C(CS(=O)(=O)O)NC(CO)(CO)CO |
Specifications
| 7365-44-8 | |
| White | |
| MFCD00007532 | |
| tes, n-tris hydroxymethyl methyl-2-aminoethanesulfonic acid, tes buffer, tes free acid, ethanesulfonic acid, 2-2-hydroxy-1,1-bis hydroxymethyl ethyl amino, 2-1,3-dihydroxy-2-hydroxymethyl propan-2-yl amino ethanesulfonic acid, 2-2-hydroxy-1,1-dihydroxymethyl-ethylamino-ethanesulfonic acid, tris hydroxymethyl methyl-2-aminomethane sulfonic acid, 2-tris hydroxymethyl methylamino ethane-1-sulphonic acid, n-tri hydroxymethyl methyltaurine | |
| C(CS(=O)(=O)O)NC(CO)(CO)CO | |
| 229.247 | |
| CHEBI:44356 | |
| ≥98.0% (T) | |
| N-Tris(hydroxymethyl)methyl-2-aminoethanesulfonic Acid [Good′s buffer component for biological |
| 216°C | |
| C6H15NO6S | |
| 25 g | |
| JOCBASBOOFNAJA-UHFFFAOYSA-N | |
| 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonic acid | |
| 81831 | |
| 229.25 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes