missing translation for 'onlineSavingsMsg'
Learn More

N-Trimethylsilylimidazole 98.0+%, TCI America™

Catalog No. p-7136348
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T058525G 25 g
T0585100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T058525G Supplier TCI America Supplier No. T058525G

Chemical Identifiers

CAS 18156-74-6
Molecular Formula C6H12N2Si
Molecular Weight (g/mol) 140.26
MDL Number MFCD00005280
InChI Key YKFRUJSEPGHZFJ-UHFFFAOYSA-N
Synonym n-trimethylsilyl imidazole, 1-trimethylsilyl imidazole, 1-trimethylsilyl-1h-imidazole, tsim, 1h-imidazole, 1-trimethylsilyl, n-trimethylsilylimidazole, trimethylsilyl imidazole, 1-trimethylsilylimidazole, n-trimethylsilyl imidazol, imidazole, 1-trimethylsilyl
PubChem CID 28925
ChEBI CHEBI:85063
IUPAC Name 1-(trimethylsilyl)-1H-imidazole
SMILES C[Si](C)(C)N1C=CN=C1

Specifications

Color Colorless
Boiling Point 91°C
Quantity 25 g
UN Number 1993
Formula Weight 140.26
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material N-Trimethylsilylimidazole [Trimethylsilylating Reagent]
RTECSNumber NI8700000
TSCA Yes
Recommended Storage Refrigerator