missing translation for 'onlineSavingsMsg'
Learn More

N-Trimethylsilylimidazole Kit SIM 1 mL *8/Reaction vial, capacity 2 mL, 98.0+%, TCI America™

Catalog No. T0693KIT
Change view
Click to view available options
Quantity:
Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0693KIT Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T0693KIT Supplier TCI America Supplier No. T06931KIT

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18156-74-6
Molecular Formula C6H12N2Si
Molecular Weight (g/mol) 140.26
MDL Number MFCD00005280
InChI Key YKFRUJSEPGHZFJ-UHFFFAOYSA-N
Synonym n-trimethylsilyl imidazole, 1-trimethylsilyl imidazole, 1-trimethylsilyl-1h-imidazole, tsim, 1h-imidazole, 1-trimethylsilyl, n-trimethylsilylimidazole, trimethylsilyl imidazole, 1-trimethylsilylimidazole, n-trimethylsilyl imidazol, imidazole, 1-trimethylsilyl
PubChem CID 28925
ChEBI CHEBI:85063
IUPAC Name 1-(trimethylsilyl)-1H-imidazole
SMILES C[Si](C)(C)N1C=CN=C1

Specifications

Color Colorless
Quantity Kit
UN Number 1993
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material N-Trimethylsilylimidazole Kit SIM 1 mL * 8 / Reaction vial, capacity 2 mL * 8 [Trimethylsilylating R
RTECSNumber NI8700000
TSCA Yes
Recommended Storage Refrigerator