missing translation for 'onlineSavingsMsg'
Learn More

N-(Trimethylsilyl)morpholine 97.0+%, TCI America™

Catalog No. T127725ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T127725ML 25 mL
T12775ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T127725ML Supplier TCI America Supplier No. T127725ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13368-42-8
Molecular Formula C7H17NOSi
Molecular Weight (g/mol) 159.304
MDL Number MFCD00010098
InChI Key JJOWIQMPCCUIGA-UHFFFAOYSA-N
Synonym 4-trimethylsilyl morpholine, n-trimethylsilyl morpholine, n-trimethylsilylmorpholine, trimethyl morpholin-4-yl silane, morpholine, 4-trimethylsilyl, trimethylsilyl morpholin, morpholinotrimethylsilane, trimethylmorpholinosilane, acmc-1btin, timtec-bb sbb009054
PubChem CID 559763
IUPAC Name trimethyl(morpholin-4-yl)silane
SMILES C[Si](C)(C)N1CCOCC1

Specifications

Color Colorless
Boiling Point 102°C
Quantity 25 mL
UN Number 2924
Formula Weight 159.30
Percent Purity ≥97.0% (GC)
Grade Reagent
Physical Form Liquid
Chemical Name or Material N-(Trimethylsilyl)morpholine
TSCA No
Recommended Storage Refrigerator