missing translation for 'onlineSavingsMsg'
Learn More

N-(Trimethylsilyl)imidazole, TMSI, EMD-Millipore

Catalog No. m1097710005
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1097710005 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1097710005 Supplier Technidata Supplier No. 1.09771.0005
Only null left

Chemical Identifiers

CAS 18156-74-6
Molecular Formula C6H12N2Si
Molecular Weight (g/mol) 140.26
MDL Number MFCD00005280
InChI Key YKFRUJSEPGHZFJ-UHFFFAOYSA-N
Synonym TSIM
PubChem CID 28925
ChEBI CHEBI:85063
IUPAC Name 1-(trimethylsilyl)-1H-imidazole
SMILES C[Si](C)(C)N1C=CN=C1

Specifications

Packaging Glass
Ignition Point 540°C DIN 51794
Viscosity 2.5mm2/sec., kinematic (25°C)
Density Density: 0.95g/cm3 (25°C)
Quantity 5 mL
Formula Weight 140.25
Chemical Name or Material N-(Trimethylsilyl)imidazole
Recommended Storage Store at 15° to 25°C