missing translation for 'onlineSavingsMsg'
Learn More

N-(Trimethylsilyl)imidazole, 97%

Catalog No. AC167131000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167131000 100 g Glass bottle
AC167130250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC167131000 Supplier Thermo Scientific Chemicals Supplier No. 167131000
Only null left

N-(Trimethylsilyl)imidazole, 97%, Quantity: 250g, Packaging: Glass bottle, Boiling Point: 93 deg.C to 94 deg.C (13.0mmHg), Colorless to Yellow, Melting Point: -42 deg.C, Molecular Weight: 140.26, Percent Purity: 97%, Assay Percent Range: 96% min. (GC), CAS: 18156-74-6 | CAS: 18156-74-6 | C6H12N2Si | 140.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Silylation reagent

Chemical Identifiers

CAS 18156-74-6
Molecular Formula C6H12N2Si
Molecular Weight (g/mol) 140.26
MDL Number MFCD00005280
InChI Key YKFRUJSEPGHZFJ-UHFFFAOYSA-N
Synonym n-trimethylsilyl imidazole, 1-trimethylsilyl imidazole, 1-trimethylsilyl-1h-imidazole, tsim, 1h-imidazole, 1-trimethylsilyl, n-trimethylsilylimidazole, trimethylsilyl imidazole, 1-trimethylsilylimidazole, n-trimethylsilyl imidazol, imidazole, 1-trimethylsilyl
PubChem CID 28925
ChEBI CHEBI:85063
IUPAC Name imidazol-1-yl(trimethyl)silane
SMILES C[Si](C)(C)N1C=CN=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -42°C
Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 93°C to 94°C (13.0 mmHg)
Flash Point 5°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4750 to 1.478
Quantity 100 g
Fieser 07,399; 11,575; 01,337; 02,177
Specific Gravity 0.95
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in most common organic solvents
Viscosity 2.5 mm2/s (25°C)
Formula Weight 140.26
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N-(Trimethylsilyl)imidazole
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective
EINECSNumber 242-040-3

RUO – Research Use Only