missing translation for 'onlineSavingsMsg'
Learn More

N-(Trimethylsilyl)diethylamine 98.0+%, TCI America™

Catalog No. t049225ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
Catalog No. Quantity
T049225ML 25 mL
1 options

Catalog No. T049225ML

Supplier: TCI America T049225ML

Only null left

Chemical Identifiers

CAS 996-50-9
Molecular Formula C7H19NSi
Molecular Weight (g/mol) 145.32
MDL Number MFCD00009040
InChI Key JOOMLFKONHCLCJ-UHFFFAOYSA-N
Synonym n,n-diethyl-1,1,1-trimethylsilylamine, trimethylsilyldiethylamine, diethylaminotrimethylsilane, n,n-diethyltrimethylsilylamine, n,n-diethyl-1,1,1-trimethylsilanamine, n,n-diethylaminotrimethylsilane, diethyl trimethylsilyl amine, n-trimethylsilyldiethylamine, diethylamino trimethylsilane, n-trimethylsilyl diethylamine
PubChem CID 70454
ChEBI CHEBI:85070
IUPAC Name diethyl(trimethylsilyl)amine
SMILES CCN(CC)[Si](C)(C)C

Specifications

Color Yellow
Boiling Point 126°C
Quantity 25 mL
UN Number 2733
Formula Weight 145.32
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(Trimethylsilyl)diethylamine