missing translation for 'onlineSavingsMsg'
Learn More

N-(Trimethylsilyl)diethylamine, 97%

Catalog No. p-7024013
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1814914 25 g
AAA1814922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1814914 Supplier Thermo Scientific Chemicals Supplier No. A1814914

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 996-50-9 | C7H19NSi | 145.32 g/mol

It is used to derivative polar organic compounds. N,N-Diethyl(trimethylsilyl)amine is a chemical reagent with wide application in organic chemistry and used in stereo specific addition reactions as well as in SN2 nucleophilic substitutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to derivative polar organic compounds. N,N-Diethyl(trimethylsilyl)amine is a chemical reagent with wide application in organic chemistry and used in stereo specific addition reactions as well as in SN2 nucleophilic substitutions.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 996-50-9
Molecular Formula C7H19NSi
Molecular Weight (g/mol) 145.32
MDL Number MFCD00009040
InChI Key JOOMLFKONHCLCJ-UHFFFAOYSA-N
Synonym n,n-diethyl-1,1,1-trimethylsilylamine, trimethylsilyldiethylamine, diethylaminotrimethylsilane, n,n-diethyltrimethylsilylamine, n,n-diethyl-1,1,1-trimethylsilanamine, n,n-diethylaminotrimethylsilane, diethyl trimethylsilyl amine, n-trimethylsilyldiethylamine, diethylamino trimethylsilane, n-trimethylsilyl diethylamine
PubChem CID 70454
ChEBI CHEBI:85070
IUPAC Name N-ethyl-N-trimethylsilylethanamine
SMILES CCN(CC)[Si](C)(C)C

Specifications

Boiling Point 125°C to 126°C
Melting Point -10°C
Quantity 25 g
UN Number UN2733
Beilstein 635718
Flash Point 10°C (50°F)
Solubility Information Hydrolyzes in water.
Formula Weight 145.32
Percent Purity 98%
Odor Amine-like
Refractive Index 1.411
Sensitivity Moisture sensitive
Density 0.76
Chemical Name or Material N-(Trimethylsilyl)diethylamine
Show More Show Less
Hazard Category H225-H314
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 213-637-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only