missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(Trimethylsilyl)diethylamine, 97%
Description
It is used to derivative polar organic compounds. N,N-Diethyl(trimethylsilyl)amine is a chemical reagent with wide application in organic chemistry and used in stereo specific addition reactions as well as in SN2 nucleophilic substitutions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used to derivative polar organic compounds. N,N-Diethyl(trimethylsilyl)amine is a chemical reagent with wide application in organic chemistry and used in stereo specific addition reactions as well as in SN2 nucleophilic substitutions.
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is used to derivative polar organic compounds. N,N-Diethyl(trimethylsilyl)amine is a chemical reagent with wide application in organic chemistry and used in stereo specific addition reactions as well as in SN2 nucleophilic substitutions.
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Boiling Point | 125°C to 126°C |
| Melting Point | -10°C |
| Quantity | 100 g |
| Flash Point | 10°C (50°F) |
| Odor | Amine-like |
| Refractive Index | 1.411 |
| Sensitivity | Moisture sensitive |
| Density | 0.76 |
| UN Number | UN2733 |
| Beilstein | 635718 |
| Show More |
Safety and Handling
| Hazard Category | H225-H314 |
| Hazard Statement | GHS H Statement H225-H314-H318 Highly flammable liquid and vapour. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S. |
| EINECSNumber | 213-637-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only