missing translation for 'onlineSavingsMsg'
Learn More

N-(Trimethylsilyl)bis(trifluoromethanesulfonyl)imide 97.0+%, TCI America™

Catalog No. p-7137198
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23921G 1 g
T23925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23921G Supplier TCI America Supplier No. T23921G
Only null left

Chemical Identifiers

CAS 82113-66-4
Molecular Formula C5H9F6NO4S2Si
Molecular Weight (g/mol) 353.325
InChI Key MLIRNWUYOYIGBZ-UHFFFAOYSA-N
Synonym [Bis(trifluoromethanesulfonyl)amino]trimethylsilane, TMS Triflimide
PubChem CID 11013624
IUPAC Name 1,1,1-trifluoro-N-(trifluoromethylsulfonyl)-N-trimethylsilylmethanesulfonamide
SMILES C[Si](C)(C)N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F

Specifications

Boiling Point 115°C
Quantity 1 g
Formula Weight 353.33
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material N-(Trimethylsilyl)bis(trifluoromethanesulfonyl)imide
TSCA No
Recommended Storage Freezer