Learn More
n-Tridecanoic acid, 97%
CAS: 638-53-9 | C13H26O2 | 214.35 g/mol
$103.68 - $103.68
Chemical Identifiers
| CAS | 638-53-9 |
|---|---|
| Molecular Formula | C13H26O2 |
| Molecular Weight (g/mol) | 214.35 |
| MDL Number | MFCD00002741 |
| InChI Key | SZHOJFHSIKHZHA-UHFFFAOYSA-N |
| Synonym | n-tridecanoic acid, tridecylic acid, n-tridecoic acid, unii-19936liy2v, anion, tridecanoic acid, wln: qv12, c10-16 fatty acids, fatty acids, c10-16, carboxylic acids, c10-16 |
| PubChem CID | 12530 |
| ChEBI | CHEBI:45919 |
| IUPAC Name | tridecanoic acid |
| SMILES | CCCCCCCCCCCCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC139520100
|
Thermo Scientific Chemicals
139520100 |
10 g | Glass bottle |
Each for $103.68
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 638-53-9 | |
| 214.35 | |
| SZHOJFHSIKHZHA-UHFFFAOYSA-N | |
| 12530 | |
| tridecanoic acid |
| C13H26O2 | |
| MFCD00002741 | |
| n-tridecanoic acid, tridecylic acid, n-tridecoic acid, unii-19936liy2v, anion, tridecanoic acid, wln: qv12, c10-16 fatty acids, fatty acids, c10-16, carboxylic acids, c10-16 | |
| CHEBI:45919 | |
| CCCCCCCCCCCCC(=O)O |
Specifications
| 638-53-9 | |
| 100.0 | |
| White to Yellow | |
| 113°C | |
| 97% | |
| C13H26O2 | |
| MFCD00002741 | |
| 02, 364 | |
| Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether | |
| CCCCCCCCCCCCC(=O)O | |
| 214.35 | |
| CHEBI:45919 | |
| 97% | |
| n-Tridecanoic acid, 97% |
| 96.0 | |
| 40°C to 45°C | |
| 236°C (100.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| CH3(CH2)11CO2H | |
| 10 g | |
| n-tridecanoic acid, tridecylic acid, n-tridecoic acid, unii-19936liy2v, anion, tridecanoic acid, wln: qv12, c10-16 fatty acids, fatty acids, c10-16, carboxylic acids, c10-16 | |
| SZHOJFHSIKHZHA-UHFFFAOYSA-N | |
| tridecanoic acid | |
| 12530 | |
| 214.35 | |
| Crystalline Chunks, Powder or Flakes |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 211-341-1
RTECSNumber : YD3850000
TSCA : TSCA
RUO – Research Use Only