missing translation for 'onlineSavingsMsg'
Learn More

n-Tridecane, 98+%

CAS: 629-50-5 | C13H28 | 184.37 g/mol

Supplier:  Thermo Scientific Chemicals A1822022

 View more versions of this product

Catalog No. AAA1822022

Only null left
Explore available promotions

Description

Description

n-Tridecane is used as a distillation chaser. It is a component of various fuels and solvents used in industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Tridecane is used as a distillation chaser. It is a component of various fuels and solvents used in industry.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

n-Tridecane
-5°C
235°C
C13H28
CH3(CH2)11CH3
100 g
n-tridecane, tridekan, alkanes, c12-14, dodecane, methyl, unii-a3lzf0l939, tridecane, n, trd, tridecane, analytical standard, tridecan, tridecyl group
IIYFAKIEWZDVMP-UHFFFAOYSA-N
tridecane
12388
184.37
629-50-5
0.755
79°C (174°F)
1.425
MFCD00008979
1733089
Immiscible with water.
CCCCCCCCCCCCC
184.37
CHEBI:35998
≥98%

Chemical Identifiers

629-50-5
184.37
IIYFAKIEWZDVMP-UHFFFAOYSA-N
12388
tridecane
C13H28
MFCD00008979
n-tridecane, tridekan, alkanes, c12-14, dodecane, methyl, unii-a3lzf0l939, tridecane, n, trd, tridecane, analytical standard, tridecan, tridecyl group
CHEBI:35998
CCCCCCCCCCCCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H304
May be fatal if swallowed and enters airways.

P210-P280-P301+P310-P331-P370+P378q-P501c

H227-H304

EINECSNumber : 211-093-4

RTECSNumber : YD3025000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only