missing translation for 'onlineSavingsMsg'
Learn More

n-Tridecane, 98+%

Catalog No. AAA1822036
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1822036 500 g
AAA1822014 25 g
AAA1822022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1822036 Supplier Thermo Scientific Chemicals Supplier No. A1822036
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 629-50-5 | C13H28 | 184.37 g/mol

n-Tridecane is used as a distillation chaser. It is a component of various fuels and solvents used in industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Tridecane is used as a distillation chaser. It is a component of various fuels and solvents used in industry.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 629-50-5
Molecular Formula C13H28
Molecular Weight (g/mol) 184.37
MDL Number MFCD00008979
InChI Key IIYFAKIEWZDVMP-UHFFFAOYSA-N
Synonym n-tridecane, tridekan, alkanes, c12-14, dodecane, methyl, unii-a3lzf0l939, tridecane, n, trd, tridecane, analytical standard, tridecan, tridecyl group
PubChem CID 12388
ChEBI CHEBI:35998
IUPAC Name tridecane
SMILES CCCCCCCCCCCCC

Specifications

Melting Point -5°C
Density 0.755
Boiling Point 235°C
Flash Point 79°C (174°F)
Refractive Index 1.425
Linear Formula CH3(CH2)11CH3
Quantity 500 g
Beilstein 1733089
Solubility Information Immiscible with water.
Formula Weight 184.37
Percent Purity ≥98%
Chemical Name or Material n-Tridecane
Show More Show Less
Hazard Category H227-H304
Hazard Statement GHS H Statement
H304
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P280-P301+P310-P331-P370+P378q-P501c
EINECSNumber 211-093-4
RTECSNumber YD3025000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only