Learn More
n-Tricosane, 99%
CAS: 638-67-5 | C23H48 | 324.637 g/mol
Supplier: Thermo Scientific Chemicals A1457714
Description
n-Tricosane is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsn-Tricosane is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions in a well sealed container. Incompatible with oxidizing agents.
Specifications
| n-Tricosane | |
| 47°C to 50°C | |
| 199°C to 200°C (3 mmHg) | |
| C23H48 | |
| MFCD00009350 | |
| 1756000 | |
| Insoluble in water. | |
| CCCCCCCCCCCCCCCCCCCCCCC | |
| 324.637 | |
| CHEBI:32934 | |
| 99% |
| 638-67-5 | |
| 0.7969 | |
| >110°C (230°F) | |
| 1.4468 | |
| 25 g | |
| n-tricosane, tricosan, docosane, methyl, unii-t166b8r1vc, ch3-ch2 21-ch3, tricosane, analytical standard, tricosane, tricosane standardmaterialforgc, unii-j3n6x3yk96 component | |
| FIGVVZUWCLSUEI-UHFFFAOYSA-N | |
| tricosane | |
| 12534 | |
| 324.64 |
Chemical Identifiers
| 638-67-5 | |
| 324.637 | |
| FIGVVZUWCLSUEI-UHFFFAOYSA-N | |
| 12534 | |
| tricosane |
| C23H48 | |
| MFCD00009350 | |
| n-tricosane, tricosan, docosane, methyl, unii-t166b8r1vc, ch3-ch2 21-ch3, tricosane, analytical standard, tricosane, tricosane standardmaterialforgc, unii-j3n6x3yk96 component | |
| CHEBI:32934 | |
| CCCCCCCCCCCCCCCCCCCCCCC |
Safety and Handling
EINECSNumber : 211-347-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only