missing translation for 'onlineSavingsMsg'
Learn More

n-Tricosane, 99%

Catalog No. AAA1457706
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1457706 5 g
AAA1457714 25 g
AAA1457722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1457706 Supplier Thermo Scientific Chemicals Supplier No. A1457706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 638-67-5 | C23H48 | 324.637 g/mol

n-Tricosane is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Tricosane is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 638-67-5
Molecular Formula C23H48
Molecular Weight (g/mol) 324.637
MDL Number MFCD00009350
InChI Key FIGVVZUWCLSUEI-UHFFFAOYSA-N
Synonym n-tricosane, tricosan, docosane, methyl, unii-t166b8r1vc, ch3-ch2 21-ch3, tricosane, analytical standard, tricosane, tricosane standardmaterialforgc, unii-j3n6x3yk96 component
PubChem CID 12534
ChEBI CHEBI:32934
IUPAC Name tricosane
SMILES CCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 47°C to 50°C
Density 0.7969
Boiling Point 199°C to 200°C (3 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4468
Quantity 5 g
Beilstein 1756000
Solubility Information Insoluble in water.
Formula Weight 324.64
Percent Purity 99%
Chemical Name or Material n-Tricosane
Show More Show Less
EINECSNumber 211-347-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only