missing translation for 'onlineSavingsMsg'
Learn More

N-TIPS pyrrole-3-boronic acid pinacol ester, 95%

Catalog No. AC441020010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC441020010 1 g Glass bottle
AC441020050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC441020010 Supplier Thermo Scientific Chemicals Supplier No. 441020010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 365564-11-0 | C19H36BNO2Si | 349.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 365564-11-0
Molecular Formula C19H36BNO2Si
Molecular Weight (g/mol) 349.40
MDL Number MFCD08063117
InChI Key GWFIZBYDIHGZRJ-UHFFFAOYSA-N
Synonym 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-tris 1-methylethyl silyl-1h-pyrrole, 3-tetramethyl-1,3,2-dioxaborolan-2-yl-1-tris propan-2-yl silyl-1h-pyrrole, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-triisopropylsilyl-1h-pyrrole, 3-pinacolateboryl-1-tips-pyrrole, 1-triisopropylsilyl-1h-pyrrol-3-ylboronic acid pinacol ester, 1-triisopropylsilyl-1h-pyrrole-3-boronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-triisopropylsilyl pyrrole, pubchem18460, pyrrole-3-boronic acid, pinacol ester;, n-tips pyrrole-3-boronic acid pinacol ester
PubChem CID 11450924
IUPAC Name tri(propan-2-yl)-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrol-1-yl]silane
SMILES CC(C)[Si](C(C)C)(C(C)C)N1C=CC(=C1)B1OC(C)(C)C(C)(C)O1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 70°C to 72°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 349.4
Percent Purity 95%
Physical Form Powder or Solid
Chemical Name or Material N-TIPS pyrrole-3-boronic acid pinacol ester
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only