missing translation for 'onlineSavingsMsg'
Learn More

N-TIPS indole-3-boronic acid pinacol ester, 97%

Catalog No. AC441030050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC441030050 5 g
AC441030010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC441030050 Supplier Thermo Scientific Chemicals Supplier No. 441030050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 476004-82-7 | C23H38BNO2Si | 399.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 476004-82-7
Molecular Formula C23H38BNO2Si
Molecular Weight (g/mol) 399.46
MDL Number MFCD16659107
InChI Key MXFFSQNKCJHCKO-UHFFFAOYSA-N
Synonym 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-triisopropylsilyl-1h-indole, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1-triisopropylsilanyl-1h-indole, n-tips indole-3-boronic acid pinacol ester, 2-1-triisopropylsilyl-1h-indole-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 11395451
IUPAC Name tri(propan-2-yl)-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indol-1-yl]silane
SMILES CC(C)[Si](C(C)C)(C(C)C)N1C=C(B2OC(C)(C)C(C)(C)O2)C2=CC=CC=C12

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 111°C to 115°C
Color Pink-White to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 399.46
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material N-TIPS indole-3-boronic acid pinacol ester
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only