missing translation for 'onlineSavingsMsg'
Learn More

N-Thionylaniline 98.0+%, TCI America™

Catalog No. T021525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T021525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T021525G Supplier TCI America Supplier No. T021525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1122-83-4
Molecular Formula C6H5NOS
Molecular Weight (g/mol) 139.172
MDL Number MFCD00002084
InChI Key FIOJWGRGPONADF-UHFFFAOYSA-N
Synonym n-thionylaniline, n-sulfinylaniline, benzenamine, n-sulfinyl, thionylaniline, phenylthionylamine, phenylsulfinylamine, sulfinylphenylamine, aniline, n-sulfinyl, thionyl imide, phenyl, sulfurous imide, phenyl
PubChem CID 70739
IUPAC Name (sulfinylamino)benzene
SMILES C1=CC=C(C=C1)N=S=O

Specifications

Color Yellow
Boiling Point 200°C
Quantity 25 g
Formula Weight 139.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Thionylaniline