missing translation for 'onlineSavingsMsg'
Learn More

n-Tetradecylbenzene, 97%

Catalog No. p-7024778
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1998806 5 g
AAA1998814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1998806 Supplier Thermo Scientific Chemicals Supplier No. A1998806
Only null left

CAS: 1459-10-5 | C20H34 | 274.492 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1459-10-5
Molecular Formula C20H34
Molecular Weight (g/mol) 274.492
MDL Number MFCD00015080
InChI Key JZALLXAUNPOCEU-UHFFFAOYSA-N
Synonym 1-phenyltetradecane, benzene, tetradecyl, 1-tetradecylbenzene, tetradecane, 1-phenyl, myristylbenzene, n-tetradecylbenzene, unii-i0y9j0o95a, phenyl-n-tetradecane, acmc-209cvz, 1-phenyltetradecane, analytical standard
PubChem CID 15086
IUPAC Name tetradecylbenzene
SMILES CCCCCCCCCCCCCCC1=CC=CC=C1

Specifications

Melting Point 9°C
Density 0.8565
Boiling Point 196°C (6 mmHg)
Refractive Index 1.4856
Quantity 5 g
Formula Weight 274.49
Percent Purity 97%
Chemical Name or Material n-Tetradecylbenzene
Hazard Category H317-H351
Precautionary Statement P201-P202-P261-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c
EINECSNumber 215-950-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only