missing translation for 'onlineSavingsMsg'
Learn More

n-Tetradecane, 99%

Catalog No. p-3346546
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1467222 100 g
AAA1467236 500 g
AAA146720E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1467222 Supplier Thermo Scientific Chemicals Supplier No. A1467222
Only null left

CAS: 629-59-4 | C14H30 | 198.394 g/mol

n-Tetradecane is used to study the thermodiffusion behavior of the microemulsion droplets of water, n-alkanes and pentaethylene glycol monododecyl ether. It is used as reference material in gas chromatographic analysis, as a solvent and finds applications in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Tetradecane is used to study the thermodiffusion behavior of the microemulsion droplets of water, n-alkanes and pentaethylene glycol monododecyl ether. It is used as reference material in gas chromatographic analysis, as a solvent and finds applications in organic synthesis.

Solubility
Soluble in ether, acetone and alcohol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 629-59-4
Molecular Formula C14H30
Molecular Weight (g/mol) 198.394
MDL Number MFCD00008986
InChI Key BGHCVCJVXZWKCC-UHFFFAOYSA-N
Synonym n-tetradecane, alkanes, c14-16, tridecane, methyl, ccris 715, tetradecane, analytical standard, alkanes, c14-30, myristyl, tetradekan, n-teradecane, olefine
PubChem CID 12389
ChEBI CHEBI:41253
IUPAC Name tetradecane
SMILES CCCCCCCCCCCCCC

Specifications

Melting Point 6°C
Density 0.7628
Boiling Point 252°C to 254°C
Flash Point 99°C (211°F)
Assay Percent Range 99%
Refractive Index 1.429
Linear Formula CH3(CH2)12CH3
Quantity 100 g
Solubility Information Soluble in ether,acetone and alcohol. Insoluble in water.
Formula Weight 198.39
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material n-Tetradecane
Show More Show Less
Hazard Category H304
Hazard Statement GHS H Statement
H304-H336
Precautionary Statement P301+P310-P331-P501c
EINECSNumber 211-096-0
RTECSNumber XB8000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only