missing translation for 'onlineSavingsMsg'
Learn More

n-Tetradecane, 99%, Spectrum™ Chemical

Catalog No. 18612054
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
20 L
200 L
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612054 500 mL Amber Glass Bottle
18612050 100 mL Amber Glass Bottle
18612051 2.5 L Amber Glass Bottle
18612052 20 L Amber Glass Bottle
18612053 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18612054 Supplier Spectrum Chemical Mfg Cor Supplier No. T2007500ML
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 629-59-4

Specifications

CAS Max % 1
Boiling Point 252°C/485.6°F
Packaging Amber Glass Bottle
Quantity 500 mL
Molecular Weight (g/mol) 198.39
Percent Purity 0.99
Grade Ungraded
Physical Form Liquid
Chemical Name or Material n-Tetradecane

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.