missing translation for 'onlineSavingsMsg'
Learn More

n-Tetracontane, 97% min

Catalog No. p-7048698
Change view
Click to view available options
Quantity:
0.5 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1127402 0.5 g
AA1127404 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1127402 Supplier Thermo Scientific Chemicals Supplier No. 01127402
Only null left

CAS: 4181-95-7 | C40H82 | 563.10 g/mol

n-Tetracontane is used in a passivation layer in copper-phthalocyanine field-effect transistors for improving the charge carrier mobility and used to modify the quasi two-dimensional surface electron system on Au(111).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4181-95-7
Molecular Formula C40H82
Molecular Weight (g/mol) 563.10
MDL Number MFCD00015267
InChI Key KUPLEGDPSCCPJI-UHFFFAOYSA-N
Synonym n-tetracontane, unii-i79s9ixb9y, alkane c40, i79s9ixb9y, tetracontane, analytical standard, acmc-209jlw, n-tetracontane 500mg, tetracontane gc
PubChem CID 20149
IUPAC Name tetracontane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 80°C to 84°C
Color White
Density 0.779 g/mL
Boiling Point 150°C (0.001 mmHg)
Linear Formula CH3(CH2)38CH3
Quantity 0.5 g
Beilstein 1800152
Solubility Information Soluble in chloroform. Slightly soluble in methanol.Insoluble in water.
Formula Weight 563.09
Percent Purity ≤97%
Physical Form Flakes
Chemical Name or Material n-Tetracontane
Show More Show Less
EINECSNumber 224-055-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only