missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butylmethylamine, 97%

Catalog No. AAH2724206
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2724206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2724206 Supplier Thermo Scientific Chemicals Supplier No. H2724206

CAS: 14610-37-8 | C5H13N | 87.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14610-37-8
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00042853
InChI Key ZQGJEUVBUVKZKS-UHFFFAOYSA-N
Synonym n-methyl-tert-butylamine, n-tert-butylmethylamine, 2-propanamine, n,2-dimethyl, tert-butylmethylamine, tert-butyl methyl amine, n-methyl-tert.-butylamine, 2-propanamine,n,2-dimethyl, t-butylmethylamine, methyl-t-butylamine, n-t-butylmethylamine
PubChem CID 84552
IUPAC Name N,2-dimethylpropan-2-amine
SMILES CC(C)(C)NC

Specifications

Density 0.727
Boiling Point 67°C to 69°C
Flash Point −18°C (0°F)
Odor Amine-like
Refractive Index 1.395
Linear Formula (CH3)3CNHCH3
Quantity 5 g
UN Number UN2733
Sensitivity Air Sensitive
Formula Weight 87.16
Percent Purity 97%
Chemical Name or Material N-tert-Butylmethylamine
Show More Show Less
Hazard Category H225-H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 238-646-2
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only