missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butylmethacrylamide (stabilized with MEHQ) 97.0+%, TCI America™

Catalog No. B111725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B111725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B111725G Supplier TCI America Supplier No. B111725G
Only null left

Chemical Identifiers

CAS 6554-73-0
Molecular Formula C8H15NO
Molecular Weight (g/mol) 141.214
MDL Number MFCD00043639
InChI Key QQZXAODFGRZKJT-UHFFFAOYSA-N
PubChem CID 81043
IUPAC Name N-tert-butyl-2-methylprop-2-enamide
SMILES CC(=C)C(=O)NC(C)(C)C

Specifications

Melting Point 58°C
Color White-Yellow
Quantity 25 g
Formula Weight 141.21
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-tert-Butylmethacrylamide (stabilized with MEHQ)