missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-tert-Butylethylamine, TCI America™

$93.20 - $323.89

Chemical Identifiers

CAS 4432-77-3
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.193
MDL Number MFCD00059367
InChI Key XQOIBQBPAXOVGP-UHFFFAOYSA-N
PubChem CID 521156
IUPAC Name N-ethyl-2-methylpropan-2-amine
SMILES CCNC(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B088825ML View Documents TCI America
B088825ML
25mL Each for $323.89
 
B08885ML View Documents TCI America
B08885ML
5mL Each for $93.20
 
Specifications

Chemical Identifiers

4432-77-3
101.193
XQOIBQBPAXOVGP-UHFFFAOYSA-N
N-ethyl-2-methylpropan-2-amine
C6H15N
MFCD00059367
521156
CCNC(C)(C)C

Specifications

89°C
25mL
C6H15N
2733
CCNC(C)(C)C
101.193
101.19
≥98.0% (GC,T)
Yellow
4432-77-3
MFCD00059367
XQOIBQBPAXOVGP-UHFFFAOYSA-N
N-ethyl-2-methylpropan-2-amine
521156
Clear Liquid at 20°C
N-tert-Butylethylamine
Safety and Handling

Safety and Handling

TSCA : No