missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butyldimethylsilyl)-N-methyl-trifluoroacetamide, 98%

Catalog No. AC221540010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC221540010 1 g Glass bottle
AC221540050 5 g Glass bottle
AC221540250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC221540010 Supplier Thermo Scientific Chemicals Supplier No. 221540010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 77377-52-7 | C9H18F3NOSi | 241.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent, Silylation reagent

Chemical Identifiers

CAS 77377-52-7
Molecular Formula C9H18F3NOSi
Molecular Weight (g/mol) 241.33
MDL Number MFCD00009671
InChI Key QRKUHYFDBWGLHJ-UHFFFAOYSA-N
Synonym mtbstfa, n-tert-butyldimethylsilyl-n-methyltrifluoroacetamide, n-methyl-n-tert-butyldimethylsilyl trifluoroacetamide, n-tert-butyldimethylsilyl-n-methyl-trifluoroacetamide, n-methyl-n-t-butyldimethylsilyl trifluoroacetamide, acetamide, n-1,1-dimethylethyl dimethylsilyl-2,2,2-trifluoro-n-methyl, n-t-butyldimethylsilyl-n-methyltrifluoroacetamide, n-t-butyldimethylsilyl-n-methyltrifluoro-acetamide
PubChem CID 2724275
ChEBI CHEBI:85060
IUPAC Name N-[tert-butyl(dimethyl)silyl]-2,2,2-trifluoro-N-methylacetamide
SMILES CC(C)(C)[Si](C)(C)N(C)C(=O)C(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0200g/mL
Boiling Point 170°C
Flash Point 52°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4010 to 1.403
Linear Formula CF3CON(CH3)Si(CH3)2C(CH3)3
Quantity 1 g
Specific Gravity 1.02
Solubility Information Solubility in water: decomposes
Formula Weight 241.33
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-(tert-Butyldimethylsilyl)-N-methyl-trifluoroacetamide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only