missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butyldiethanolamine 97.0+%, TCI America™

Catalog No. p-7118910
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B119425ML 25 mL
B1194500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B119425ML Supplier TCI America Supplier No. B119425ML

Chemical Identifiers

CAS 2160-93-2
Molecular Formula C8H19NO2
Molecular Weight (g/mol) 161.245
MDL Number MFCD00020584
InChI Key XHJGXOOOMKCJPP-UHFFFAOYSA-N
PubChem CID 62430
IUPAC Name 2-[tert-butyl(2-hydroxyethyl)amino]ethanol
SMILES CC(C)(C)N(CCO)CCO

Specifications

Melting Point 49°C
Boiling Point 161°C
Quantity 25 mL
UN Number 3259
Synonym N-Bis(2-hydroxyethyl)-tert-butylamine, 2,2′C-(tert-Butylimino)diethanol
Formula Weight 161.25
Percent Purity ≥97.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material N-tert-Butyldiethanolamine
EINECSNumber (2)-0353
RTECSNumber KK7178750
TSCA Yes