missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butylcarbamoyl-L-tert-leucine 98.0+%, TCI America™

Catalog No. B53241G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B53241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B53241G Supplier TCI America Supplier No. B53241G
Only null left

Chemical Identifiers

CAS 101968-85-8
Molecular Formula C11H22N2O3
Molecular Weight (g/mol) 230.308
MDL Number MFCD12796012
InChI Key RAAPXVRHYBAJQU-SSDOTTSWSA-N
Synonym (S)-2-[3-(tert-Butyl)ureido]-3,3-dimethylbutyric Acid
PubChem CID 23659740
IUPAC Name (2S)-2-(tert-butylcarbamoylamino)-3,3-dimethylbutanoic acid
SMILES CC(C)(C)C(C(=O)O)NC(=O)NC(C)(C)C

Specifications

Melting Point 182°C
Color White
Quantity 1 g
Formula Weight 230.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-tert-Butylcarbamoyl-L-tert-leucine