missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butylacrylamide 98.0+%, TCI America™

Catalog No. B070325G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B070325G 25 g
B0703100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B070325G Supplier TCI America Supplier No. B070325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 107-58-4
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.19
MDL Number MFCD00026271
InChI Key XFHJDMUEHUHAJW-UHFFFAOYSA-N
Synonym n-tert-butylacrylamide, tert-butylacrylamide, n-tert-butyl acrylamide, 2-propenamide, n-1,1-dimethylethyl, acrylamide, n-tert-butyl, n-t-butylacrylamide, unii-xj13fsh48k, n-1,1-dimethylethyl-2-propenamide, xj13fsh48k, n-tert-butyl prop-2-enamide
PubChem CID 7877
IUPAC Name N-tert-butylprop-2-enamide
SMILES CC(C)(C)NC(=O)C=C

Specifications

Melting Point 130°C
Color White
Quantity 25 g
Formula Weight 127.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-tert-Butylacrylamide
EINECSNumber (2)-1015
RTECSNumber AS3460000
TSCA Yes