Learn More
Thermo Scientific Chemicals N-tert-Butylacrylamide, 97%, pure
CAS: 107-58-4 | C7H13NO | 127.19 g/mol
$1213.34 - $1213.34
Chemical Identifiers
| CAS | 107-58-4 |
|---|---|
| Molecular Formula | C7H13NO |
| Molecular Weight (g/mol) | 127.19 |
| MDL Number | MFCD00026271 |
| InChI Key | XFHJDMUEHUHAJW-UHFFFAOYSA-N |
| Synonym | n-tert-butylacrylamide, tert-butylacrylamide, n-tert-butyl acrylamide, 2-propenamide, n-1,1-dimethylethyl, acrylamide, n-tert-butyl, n-t-butylacrylamide, unii-xj13fsh48k, n-1,1-dimethylethyl-2-propenamide, xj13fsh48k, n-tert-butyl prop-2-enamide |
| PubChem CID | 7877 |
| IUPAC Name | N-tert-butylprop-2-enamide |
| SMILES | CC(C)(C)NC(=O)C=C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC403455000
|
Thermo Scientific Chemicals
403455000 |
500 g | Glass bottle |
Each for $1,213.34
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 107-58-4 | |
| 127.19 | |
| XFHJDMUEHUHAJW-UHFFFAOYSA-N | |
| 7877 | |
| CC(C)(C)NC(=O)C=C |
| C7H13NO | |
| MFCD00026271 | |
| n-tert-butylacrylamide, tert-butylacrylamide, n-tert-butyl acrylamide, 2-propenamide, n-1,1-dimethylethyl, acrylamide, n-tert-butyl, n-t-butylacrylamide, unii-xj13fsh48k, n-1,1-dimethylethyl-2-propenamide, xj13fsh48k, n-tert-butyl prop-2-enamide | |
| N-tert-butylprop-2-enamide |
Specifications
| 107-58-4 | |
| White | |
| 96% min. (GC) | |
| C7H13NO | |
| MFCD00026271 | |
| n-tert-butylacrylamide, tert-butylacrylamide, n-tert-butyl acrylamide, 2-propenamide, n-1,1-dimethylethyl, acrylamide, n-tert-butyl, n-t-butylacrylamide, unii-xj13fsh48k, n-1,1-dimethylethyl-2-propenamide, xj13fsh48k, n-tert-butyl prop-2-enamide | |
| CC(C)(C)NC(=O)C=C | |
| 127.19 | |
| 127.19 | |
| Pure | |
| N-tert-Butylacrylamide |
| 128°C to 130°C | |
| Authentic | |
| Glass bottle | |
| CH2CHCONHC(CH3)3 | |
| 500 g | |
| XFHJDMUEHUHAJW-UHFFFAOYSA-N | |
| N-tert-butylprop-2-enamide | |
| 7877 | |
| 97% | |
| Chunks and/or Powder |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 203-505-6
RTECSNumber : AS3460000
TSCA : TSCA
RUO – Research Use Only