missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butylacetamide, 98%

Catalog No. p-7044858
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0850509 10 g
AAL0850518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0850509 Supplier Thermo Scientific Chemicals Supplier No. L0850509

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 762-84-5 | C6H13NO | 115.176 g/mol

It finds its application in the synthesis of amides using the Ritter reaction with bismuth triflate catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the synthesis of amides using the Ritter reaction with bismuth triflate catalysis.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 762-84-5
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00038244
InChI Key ACYFWRHALJTSCF-UHFFFAOYSA-N
Synonym n-tert-butyl acetamide, tert-butylacetamide, n-t-butylacetamide, acetamide, n-1,1-dimethylethyl, ch3c o nhc ch3 3, acetamide,n-1,1-dimethylethyl
PubChem CID 12985
IUPAC Name N-tert-butylacetamide
SMILES CC(=O)NC(C)(C)C

Specifications

Melting Point 95°C to 99°C
Quantity 10 g
Beilstein 1699507
Solubility Information Soluble in water.
Formula Weight 115.18
Percent Purity 98%
Chemical Name or Material N-tert-Butylacetamide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only