missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butyl-α-phenylnitrone, ≥99.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013961
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013961 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013961 Supplier Merck Emd Millipore Supplier No. 801261G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00008799
Synonym N-Benzylidene-tert-butylamine N-oxide; PBN; Phenyl N-t-butylnitrone

Specifications

Melting Point 72°C to 74°C; 73°C to 74°C
CAS 3376-24-7
Linear Formula C6H5 CH=N(O)C(CH3)3
Quantity 1 g
Solubility Information Chloroform: 50 mg/mL, clear, colorless
Percent Purity ≥99.5% (HPLC)
Grade For ESR-spectroscopy
RTECSNumber TX1760000