missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butyl-alpha-phenylnitrone 98.0+%, TCI America™

$130.92 - $419.69

Chemical Identifiers

CAS 3376-24-7
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD00008799
InChI Key IYSYLWYGCWTJSG-FMIVXFBMSA-N
Synonym e-n-benzylidene-2-methylpropan-2-amine oxide
PubChem CID 10313352
IUPAC Name N-tert-butyl-1-phenylmethanimine oxide
SMILES CC(C)(C)[N+](=CC1=CC=CC=C1)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B17011G
View Documents
TCI America
B17011G
1 g
Each for $130.92
Only null left
 
B17015G
View Documents
TCI America
B17015G
5 g
Each for $419.69
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3376-24-7
177.247
IYSYLWYGCWTJSG-FMIVXFBMSA-N
10313352
CC(C)(C)[N+](=CC1=CC=CC=C1)[O-]
C11H15NO
MFCD00008799
e-n-benzylidene-2-methylpropan-2-amine oxide
N-tert-butyl-1-phenylmethanimine oxide

Specifications

3376-24-7
C11H15NO
1 g
IYSYLWYGCWTJSG-FMIVXFBMSA-N
N-tert-butyl-1-phenylmethanimine oxide
10313352
≥98.0% (HPLC,T)
N-tert-Butyl-alpha-phenylnitrone
74°C
MFCD00008799
e-n-benzylidene-2-methylpropan-2-amine oxide
CC(C)(C)[N+](=CC1=CC=CC=C1)[O-]
177.247
177.25
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents