missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butyl-2-thiophenesulfonamide 98.0+%, TCI America™

Catalog No. p-7123476
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43301G 1 g
B43305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43301G Supplier TCI America Supplier No. B43301G
Only null left

Chemical Identifiers

CAS 100342-30-1
Molecular Formula C8H13NO2S2
Molecular Weight (g/mol) 219.317
MDL Number MFCD02047252
InChI Key CLKMBGGZGFULOO-UHFFFAOYSA-N
Synonym 2-(tert-Butylaminosulfonyl)thiophene
PubChem CID 765814
IUPAC Name N-tert-butylthiophene-2-sulfonamide
SMILES CC(C)(C)NS(=O)(=O)C1=CC=CS1

Specifications

Melting Point 84°C
Color White
Quantity 1 g
Formula Weight 219.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-tert-Butyl-2-thiophenesulfonamide