missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyloxy)succinimide, 98+%

Catalog No. p-7036232
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5449303 1 g
AAH5449306 5 g
AAH5449314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5449303 Supplier Thermo Scientific Chemicals Supplier No. H5449303
Only null left

CAS: 13139-12-3 | C9H13NO5 | 215.205 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13139-12-3
Molecular Formula C9H13NO5
Molecular Weight (g/mol) 215.205
MDL Number MFCD00037903
InChI Key VTGFSVGZCYYHLO-UHFFFAOYSA-N
Synonym boc-osu, tert-butyl 2,5-dioxopyrrolidin-1-yl carbonate, tert-butyl n-succinimidyl carbonate, n-tert-butoxycarbonyloxy succinimide, tert-butyl succinimido carbonate, 1-tert-butoxycarbonyl oxy pyrrolidine-2,5-dione, 2,5-pyrrolidinedione, 1-1,1-dimethylethoxy carbonyl oxy, carbonic acid, 1,1-dimethylethyl 2,5-dioxo-1-pyrrolidinyl ester, boc-osu, tert-butyl n-succinimidyl carbonate, n-tert-butoxycarbonyloxy succinimide boc-osu
PubChem CID 83168
IUPAC Name tert-butyl (2,5-dioxopyrrolidin-1-yl) carbonate
SMILES CC(C)(C)OC(=O)ON1C(=O)CCC1=O

Specifications

Melting Point ∽95°C (decomposition)
Quantity 1 g
Sensitivity Moisture sensitive
Formula Weight 215.2
Percent Purity ≥98%
Chemical Name or Material N-(tert-Butoxycarbonyloxy)succinimide
Hazard Category H302-H317
Hazard Statement GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
EINECSNumber 236-071-1
TSCA Yes
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only