missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyloxy)succinimide, 97%

Catalog No. AC436240050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC436240050 5 g
AC436240250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC436240050 Supplier Thermo Scientific Chemicals Supplier No. 436240050

CAS: 13139-12-3 | C9H13NO5 | 215.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13139-12-3
Molecular Formula C9H13NO5
Molecular Weight (g/mol) 215.21
InChI Key VTGFSVGZCYYHLO-UHFFFAOYSA-N
Synonym boc-osu, tert-butyl 2,5-dioxopyrrolidin-1-yl carbonate, tert-butyl n-succinimidyl carbonate, n-tert-butoxycarbonyloxy succinimide, tert-butyl succinimido carbonate, 1-tert-butoxycarbonyl oxy pyrrolidine-2,5-dione, 2,5-pyrrolidinedione, 1-1,1-dimethylethoxy carbonyl oxy, carbonic acid, 1,1-dimethylethyl 2,5-dioxo-1-pyrrolidinyl ester, boc-osu, tert-butyl n-succinimidyl carbonate, n-tert-butoxycarbonyloxy succinimide boc-osu
PubChem CID 83168
IUPAC Name tert-butyl (2,5-dioxopyrrolidin-1-yl) carbonate
SMILES CC(C)(C)OC(=O)ON1C(=O)CCC1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 88°C to 91°C
Color White
Elemental Analysis 48.22 to 52.24% C, 6.25 to 6.77% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 5 g
Formula Weight 215.21
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-(tert-Butoxycarbonyloxy)succinimide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 236-071-1

RUO – Research Use Only