missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-proline Methyl Ester 95.0+%, TCI America™

Catalog No. B38431G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38431G 1 g
B38435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38431G Supplier TCI America Supplier No. B38431G

Chemical Identifiers

CAS 74844-91-0
Molecular Formula C11H19NO5
Molecular Weight (g/mol) 245.28
MDL Number MFCD00076981
InChI Key MZMNEDXVUJLQAF-SFYZADRCSA-N
Synonym boc-hyp-ome, n-boc-trans-4-hydroxy-l-proline methyl ester, 2s,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2s,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, boc-l-hydroxyproline methyl ester, boc-hydroxyproline-ome, methyl trans-1-boc-4-hydroxy-l-prolinate, n-tert-butoxycarbonyl-trans-4-hydroxy-l-proline methyl ester, trans-1-tert-butoxycarbonyl-4-hydroxy-l-proline methyl ester, n-boc-trans-4-hydroxyl-l-proline methyl ester
PubChem CID 2734883
IUPAC Name 1-tert-butyl 2-methyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate
SMILES COC(=O)[C@@H]1C[C@@H](O)CN1C(=O)OC(C)(C)C

Specifications

Melting Point 96°C
Color White
Boiling Point 132°C
Quantity 1 g
Formula Weight 245.28
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-proline Methyl Ester