missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-O-tert-butyl-L-tyrosine 98.0+%, TCI America™

$54.06 - $160.30

Chemical Identifiers

CAS 47375-34-8
Molecular Formula C18H27NO5
Molecular Weight (g/mol) 337.42
MDL Number MFCD00065598
InChI Key ZEQLLMOXFVKKCN-YQTOOIBONA-N
Synonym boc-tyr tbu-oh, boc-o-tert-butyl-l-tyrosine, boc-tyr tbul-oh, s-3-4-tert-butoxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, n-tert-butoxycarbonyl-o-tert-butyl-l-tyrosine, l-tyrosine, n-1,1-dimethylethoxy carbonyl-o-1,1-dimethylethyl, 2s-3-4-tert-butoxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-2-tert-butoxy carbonylamino-3-4-tert-butoxy phenyl propanoic acid, boc-tyr but-oh, pubchem12288
PubChem CID 7017901
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-[4-(tert-butoxy)phenyl]propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(OC(C)(C)C)C=C1)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B34851G
View Documents
TCI America
B34851G
1 g N/A
Only null left
 
B34855G
View Documents
TCI America
B34855G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

47375-34-8
337.42
ZEQLLMOXFVKKCN-YQTOOIBONA-N
7017901
CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(OC(C)(C)C)C=C1)C(O)=O
C18H27NO5
MFCD00065598
boc-tyr tbu-oh, boc-o-tert-butyl-l-tyrosine, boc-tyr tbul-oh, s-3-4-tert-butoxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, n-tert-butoxycarbonyl-o-tert-butyl-l-tyrosine, l-tyrosine, n-1,1-dimethylethoxy carbonyl-o-1,1-dimethylethyl, 2s-3-4-tert-butoxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-2-tert-butoxy carbonylamino-3-4-tert-butoxy phenyl propanoic acid, boc-tyr but-oh, pubchem12288
(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-[4-(tert-butoxy)phenyl]propanoic acid

Specifications

47375-34-8
White
MFCD00065598
boc-tyr tbu-oh, boc-o-tert-butyl-l-tyrosine, boc-tyr tbul-oh, s-3-4-tert-butoxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, n-tert-butoxycarbonyl-o-tert-butyl-l-tyrosine, l-tyrosine, n-1,1-dimethylethoxy carbonyl-o-1,1-dimethylethyl, 2s-3-4-tert-butoxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-2-tert-butoxy carbonylamino-3-4-tert-butoxy phenyl propanoic acid, boc-tyr but-oh, pubchem12288
CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(OC(C)(C)C)C=C1)C(O)=O
337.42
337.42
Crystalline Powder
117°C
C18H27NO5
1 g
ZEQLLMOXFVKKCN-YQTOOIBONA-N
(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-[4-(tert-butoxy)phenyl]propanoic acid
7017901
≥98.0% (HPLC,T)
N-(tert-Butoxycarbonyl)-O-tert-butyl-L-tyrosine
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'