missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-N'-methylethylenediamine 98.0+%, TCI America™
$82.79 - $246.43
Chemical Identifiers
| CAS | 122734-32-1 |
|---|---|
| Molecular Formula | C8H18N2O2 |
| Molecular Weight (g/mol) | 174.244 |
| MDL Number | MFCD06808585 |
| InChI Key | GKWGBMHXVRSFRT-UHFFFAOYSA-N |
| PubChem CID | 3435744 |
| IUPAC Name | tert-butyl N-[2-(methylamino)ethyl]carbamate |
| SMILES | CC(C)(C)OC(=O)NCCNC |
Chemical Identifiers
| 122734-32-1 | |
| 174.244 | |
| GKWGBMHXVRSFRT-UHFFFAOYSA-N | |
| tert-butyl N-[2-(methylamino)ethyl]carbamate |
| C8H18N2O2 | |
| MFCD06808585 | |
| 3435744 | |
| CC(C)(C)OC(=O)NCCNC |
Specifications
| 122734-32-1 | |
| C8H18N2O2 | |
| N-(tert-Butoxycarbonyl)-N′C-methyl-1,2-diaminoethane, N-(2-Methylaminoethyl)carbamic Acid tert-Butyl Ester, N-Boc-N′C-methyl-1,2-diaminoethane, N-Boc-N′C-methylethylenediamine, tert-Butyl N-(2-Methylaminoethyl)carbamate | |
| CC(C)(C)OC(=O)NCCNC | |
| 1 g | |
| 3435744 | |
| ≥98.0% (T) | |
| N-(tert-Butoxycarbonyl)-N′-methylethylenediamine |
| White-Yellow | |
| MFCD06808585 | |
| GKWGBMHXVRSFRT-UHFFFAOYSA-N | |
| tert-butyl N-[2-(methylamino)ethyl]carbamate | |
| 174.244 | |
| 174.24 | |
| Crystal/Lump |
Safety and Handling
TSCA : No