missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-N-methyl-L-alanine 98.0+%, TCI America™

Catalog No. B36515G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36515G 5 g
B365125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36515G Supplier TCI America Supplier No. B36515G

Chemical Identifiers

CAS 16948-16-6
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.24
MDL Number MFCD00037242
InChI Key VLHQXRIIQSTJCQ-LURJTMIESA-N
Synonym boc-n-me-ala-oh, boc-n-methyl-l-alanine, n-boc-n-methyl-l-alanine, s-2-tert-butoxycarbonyl methyl amino propanoic acid, n-tert-butoxycarbonyl-n-methyl-l-alanine, boc-n-a-methyl-l-alanine, s-2-tert-butoxycarbonyl-methyl-amino-propionic acid, l-alanine, n-1,1-dimethylethoxy carbonyl-n-methyl, 2s-2-tert-butoxycarbonyl methyl amino propanoic acid, 2s-2-tert-butoxy carbonyl methyl amino propanoic acid
PubChem CID 7009590
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl](methyl)amino}propanoic acid
SMILES C[C@H](N(C)C(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 93°C
Color White
Quantity 5 g
Formula Weight 203.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-N-methyl-L-alanine