missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-N-methyl-D-phenylalanine 98.0+%, TCI America™
$115.51 - $350.38
Chemical Identifiers
| CAS | 85466-66-6 |
|---|---|
| Molecular Formula | C15H21NO4 |
| Molecular Weight (g/mol) | 279.34 |
| MDL Number | MFCD00151890 |
| InChI Key | AJGJINVEYVTDNH-UHFFFAOYNA-N |
| Synonym | boc-n-me-d-phe-oh, boc-d-mephe-oh, n-boc-n-methyl-d-phenylalanine, n-tert-butoxycarbonyl-n-methyl-d-phenylalanine, n-methyl-boc-d-phenylalanine, r-2-tert-butoxycarbonyl methyl amino-3-phenylpropanoic acid, 2r-2-n-tert-butoxycarbonyl-n-methylamino-3-phenylpropionic acid, boc-d-me-phe-oh, ambotzbaa1044, 2r-2-tert-butoxycarbonyl methyl amino-3-phenylpropanoic acid |
| PubChem CID | 7015127 |
| IUPAC Name | 2-{[(tert-butoxy)carbonyl](methyl)amino}-3-phenylpropanoic acid |
| SMILES | CN(C(CC1=CC=CC=C1)C(O)=O)C(=O)OC(C)(C)C |
Chemical Identifiers
| 85466-66-6 | |
| 279.34 | |
| AJGJINVEYVTDNH-UHFFFAOYNA-N | |
| 7015127 | |
| CN(C(CC1=CC=CC=C1)C(O)=O)C(=O)OC(C)(C)C |
| C15H21NO4 | |
| MFCD00151890 | |
| boc-n-me-d-phe-oh, boc-d-mephe-oh, n-boc-n-methyl-d-phenylalanine, n-tert-butoxycarbonyl-n-methyl-d-phenylalanine, n-methyl-boc-d-phenylalanine, r-2-tert-butoxycarbonyl methyl amino-3-phenylpropanoic acid, 2r-2-n-tert-butoxycarbonyl-n-methylamino-3-phenylpropionic acid, boc-d-me-phe-oh, ambotzbaa1044, 2r-2-tert-butoxycarbonyl methyl amino-3-phenylpropanoic acid | |
| 2-{[(tert-butoxy)carbonyl](methyl)amino}-3-phenylpropanoic acid |
Specifications
| 85466-66-6 | |
| White-Yellow | |
| MFCD00151890 | |
| boc-n-me-d-phe-oh, boc-d-mephe-oh, n-boc-n-methyl-d-phenylalanine, n-tert-butoxycarbonyl-n-methyl-d-phenylalanine, n-methyl-boc-d-phenylalanine, r-2-tert-butoxycarbonyl methyl amino-3-phenylpropanoic acid, 2r-2-n-tert-butoxycarbonyl-n-methylamino-3-phenylpropionic acid, boc-d-me-phe-oh, ambotzbaa1044, 2r-2-tert-butoxycarbonyl methyl amino-3-phenylpropanoic acid | |
| CN(C(CC1=CC=CC=C1)C(O)=O)C(=O)OC(C)(C)C | |
| 279.34 | |
| 279.34 | |
| Crystalline Powder |
| 86°C | |
| C15H21NO4 | |
| 1 g | |
| AJGJINVEYVTDNH-UHFFFAOYNA-N | |
| 2-{[(tert-butoxy)carbonyl](methyl)amino}-3-phenylpropanoic acid | |
| 7015127 | |
| ≥98.0% (HPLC,T) | |
| N-(tert-Butoxycarbonyl)-N-methyl-D-phenylalanine |
Safety and Handling
TSCA : No